Learn More
QC Standard 21 (Primary): As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn @ 100 μg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid, VHG™
Product Code. 30003124
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003124 500 mL 500mL
30003086 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30003124 Supplier VHG Labs Supplier No. VHGQC21500
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
QC Standard 21 (Primary): As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn @ 100 μg/mL in 5% HNO3, tr. F-, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Molybdenum/Cadmium/Antimony/Selenium/Strontium/Lithium/Beryllium/Chromium/Lead/Nickel/Thallium/Zinc/Arsenic/Calcium/Copper/Iron/Magnesium/Manganese/Vanadium/Cobalt/Titanium
CAS 7439-98-7, 7440-43-9, 7440-36-0, 7782-49-2, 7440-24-6, 7439-93-2
Methods ICP/ICP-MS
Analyte or Component Names Molybdenum/Cadmium/Antimony/Selenium/Strontium/Lithium/Beryllium/Chromium/Lead/Nickel/Thallium/Zinc/Arsenic/Calcium/Copper/Iron/Magnesium/Manganese/Vanadium/Cobalt/Titanium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Molybdenum 99.95 μg/mL/Cadmium 99.96 μg/mL/Antimony 99.98 μg/mL/Selenium 99.98 μg/mL/Strontium 99.98 μg/mL/Lithium 99.99 μg/mL/Beryllium 100 μg/mL/Chromium 100 μg/mL/Lead 100 μg/mL/Nickel 100 μg/mL/Thallium 100 μg/mL/Zinc 100 μg/mL/Arsenic 100.1 μg/mL/Calcium 100.1 μg/mL/Copper 100.1 μg/mL/Iron 100.1 μg/mL/Magnesium 100.1 μg/mL/Manganese 100.1 μg/mL/Vanadium 100.1 μg/mL/Cobalt 100.2 μg/mL/Titanium 100.2 μg/mL