missing translation for 'onlineSavingsMsg'
Learn More

QC Standard 7: Si @ 50; Ag, Al, B, Ba, Na @ 100; K @ 1000 μg/mL in 5% HNO3, tr. F-, VHG™

Product Code. 30002929
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002929 500 mL 500mL
17998739 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30002929 Supplier VHG Labs Supplier No. VHGQC7500

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

QC Standard 7: Si @ 50; Ag, Al, B, Ba, Na @ 100; K @ 1000 μg/mL in 5% HNO3, tr. F-
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silicon/Aluminium/Barium/Boron/Silver/Sodium/Potassium
CAS 7440-21-3, 7429-90-5, 7440-39-3, 7440-42-8, 7440-22-4, 7440-23-5
Methods ICP/ICP-MS
Analyte or Component Names Silicon/Aluminium/Barium/Boron/Silver/Sodium/Potassium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Silicon 50 μg/mL/Aluminium 100 μg/mL/Barium 100 μg/mL/Boron 100 μg/mL/Silver 100 μg/mL/Sodium 100 μg/mL/Potassium 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.