missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 655 Biotin Conjugate Kit
Description
The biotin-labeled Qdot™ 655 nanocrystals are available for detecting streptavidin probes or for creating noncovalent conjugates with streptavidin-labeled molecules or with other biotinylated molecules using a streptavidin bridge. The product is provided as 250μL of a 2μM solution and includes 30 mL of Qdot™ incubation buffer.
Specifications
Specifications
| Quantity | 1 kit |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Room Temperature |
| Product Line | Qdot |
| Product Type | Biotin Qdot Conjugation Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?