missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Qdot™ 705 ITK™ Streptavidin Conjugate Kit
Description
The Qdot™ 705 ITK™ Streptavidin Conjugate Kit provides the Qdot™ streptavidin conjugate as 250μL of a 2μM solution. Unlike the Qdot™ streptavidin conjugates, the Qdot™ ITK™ streptavidin conjugates have the streptavidin covalently attached to the inner amphiphilic coating without a PEG linker. These Qdo™ conjugates can be used for binding biotinylated probes. The kit also includes 30 mL of Qdot™ incubation buffer.
Specifications
Specifications
| Concentration | 2 μM |
| Conjugate | Qdot 705 |
| Quantity | 1 kit |
| Content And Storage | Store in refrigerator (2–8°C). |
| Shipping Condition | Room Temperature |
| Product Line | ITK, Qdot |
| Product Type | Streptavidin Conjugate (fluorescent) |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?