Learn More
Izon qEV10 Column
Product Code. 15938090
Encompass_Preferred
Change view
Click to view available options
For use with EV/exsosome isolation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15938090 For use with EV/exsosome isolation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15938090 Supplier Izon Supplier No. SP3
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
qEV provides the most rapid and effective EV/exsosome isolation system available. qEV is based on Size Exclusion Chromatography (SEC), and takes around 15 minutes to get a pure sample of intact exosomes. The resulting samples are consistent, standardisable and repeatable.
  • The qEV10 / 70nm is designed for separating exosomes from a 10mL biological sample and comes with Luer Lock fittings. They will be of particular interest if you are working with larger fluid volumes. The Luer Lock fittings also allow for many different configurations
Specifications
Description Izon qEV10
Column Type Size Exclusion Column
For Use With (Application) EV/exsosome isolation