Learn More
ValidR BV Qualification IQ-OQ-PQ
Description
- IQ/OQ/PQ refers to the 3 activities that must be performed on equipment and machines as part of the validation of manufacturing processes: Installation Qualification (IQ), Operational Qualification (OQ), and Performance Qualification (PQ).
- Evereything is installed correctly - IQ.
- Everything is operating correctly, and that operating limits are understood - OQ.
- It operates the way in which it is intended under load - PQ
Specifications
Specifications
| For Use With (Equipment) | Labconco PSM and BSC Cabinets |
| Product Type | IQ/OQ Documentation |