Learn More
Invitrogen™ QuantiGene™ Sample Processing Kit, cultured cells
Description
This QuantiGene Sample Processing Kit for cultured cells contains reagents and instructions for the preparation of cell lysates for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA targets and QuantiGene Plex DNA assays for DNA targets.
- Easy to follow instructions
- Preserves the integrity of the RNA/DNA without degradation
- Choose the method that matches your cultured cell sample
What you get
The QuantiGene Sample Processing Kit for cultured cells contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing cell lysates from cultured cells.
Success in cell lysis
Key factors that influence the complete lysis of the cells include:
- The lysis mixture
- Proteinase K
- Vortexing or pipetting of the sample
- Storage of the lysates at -80°C
- Incubation at 50°C
The protocols provided with the QuantiGene Sample Processing Kit for cultured cells use a combination of these factors to ensure complete lysis of the samples. In addition, the lysis mixture provides the proper ionic conditions for the overnight hybridization in the QuantiGene assays.
Specifications
Specifications
| Content And Storage | • 75 mL Lysis Mixture, store at 15°C to 30°C • 750 μL Proteinase K (50 μg/μL), store at -20°C |
| Format | Kit |
| Sample Type | Cell Cultures |
| Product Line | QuantiGene |
| Product Type | Sample Processing Kit |
| Quantity | 10 Plates |
| Shipping Condition | Blue Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.