Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1063 for Human Nectin-4
Product Code. 18771029
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18771029 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18771029 Supplier R&D Systems™ Supplier No. QC133
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / Pack of 1
    Save 23.48€
    10% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with catalog number DNEC40
Quantikine Immunoassay Control Set 1063 is designed for use as quantitative controls for the determination of Nectin-4 concentrations in biological fluids.
  • Applications: ELISA, Control
  • Species Reactivity: Human
  • Format: Lyophilized
  • Composition: Recombinant human cytokines in diluted porcine serum with preservatives
  • Storage: Store at 2°C to 8°C
  • Stability: At least 6 months from date of receipt when stored as directed
  • Compatibility: For use with R&D Systems Quantikine kits, catalog number DNEC40
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Nectin-4
Sample Type Biological Fluids
Type Immunoassay Control
Gene ID (Entrez) 81607
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human
Form Lyophilized