missing translation for 'onlineSavingsMsg'
Learn More

Quartz rod, fused, 10.0mm (0.394in) dia

Product Code. 11364338
Change view
Click to view available options
Quantity:
12 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364338 12 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11364338 Supplier Thermo Scientific Alfa Aesar Supplier No. 042294.NK
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14808-60-7 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
Physical Form Rod
Quantity 12 in.
Packaging Tube
Chemical Name or Material Quartz rod

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.