Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Product Code. 10683331 Shop All DWK Life Sciences Products
Click to view available options
Size:
DN 100
DN 120
DN 150
DN 200
DN 60
Unit Size:
Each
Product Code. 10683331
Brand: DWK Life Sciences™ 290715108
Only null left
Add to basket
Add to basket
The quick release clamp is used to close flat flange vessels.
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
  • With retaining clip
  • Quick closing with quick-release fastener
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Material Stainless Steel
For Use With (Equipment) Quick Release Clamp for Closure
For Use With (Application) Quick release clamp for closure; Accessories for flat flange vessels
Size DN 120