Learn More
Quickfit™ Borosilicate Glass Inland Revenue Condenser
Product Code. 15537760
Change view
Click to view available options
Standard Taper Joint No:
19/26
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
15537760 19/26 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15537760 Supplier Quickfit Supplier No. C6/12/SC
  • ONLINE EXCLUSIVE PRICE
    553.35€
    461.01€ / Pack of 2
    Save 92.34€
    16% Off
Estimated Shipment: 11-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Inland Revenue Condenser is highly efficient. Vapour and distillate move down through the coil while coolant water flows outside the coil. Complete with detachable plastic hose connectors for the easy and safe attachment of flexible tubing.
  • Inland Revenue Pattern
  • Highly efficient
  • Vapour and distillate move down through the coil
  • Coolant water flows outside the coil
  • Complete with detachable plastic hose connectors for the easy and safe attachment of flexible tubing
Specifications
Description Surface Area X 102m2: 2.5
Length (Metric) Jacket 205 mm
Standard Taper Joint No 19/26
Material Borosilicate Glass
Type Inland Revenue Condenser