Learn More
Quickfit™ Borosilicate Glass Reversible Condenser Coil
Product Code. 15547780
Change view
Click to view available options
Standard Taper Joint No:
19/26 and 24/29
24/29 and 34/35
Length (Metric) Jacket:
315 mm (Overall) 160 mm (Effective)
335 mm (Overall) 160 mm (Effective)
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket unitSize
15547780 19/26 and 24/29 315 mm (Overall) 160 mm (Effective) Pack of 2
15557780 24/29 and 34/35 335 mm (Overall) 160 mm (Effective) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15547780 Supplier Quickfit Supplier No. CX3/23/SC
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Reversible Condenser Coil is fitted on each end with a different size cone to reduce the need for adapters. Includes detachable plastic connectors for the attachment of flexible tubing.
  • Highly versatile reflux condenser
  • Each end of the condenser is fitted with a different size cone thus reducing the need for adapters
  • With detachable plastic connectors for the attachment of flexible tubing
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) Jacket 315 mm (Overall) 160 mm (Effective)
Standard Taper Joint No 19/26 and 24/29
Material Borosilicate Glass
Type Condenser Coil