Learn More
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Product Code. 15557940
Change view
Click to view available options
For Use With (Equipment):
For model MF29/23/250 Dreschel bottle
For model MF29/3/125 Dreschel bottle
For model MF29/3/250 Dreschel bottle
For model MF29/3/500 Dreschel bottle
Description:
Stem Length: 132 mm
Stem Length: 165 mm
Stem length: 217 mm
Porosity:
0
1
2
3
Unit Size:
Each
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Description Porosity unitSize
15557940 For model MF29/3/250 Dreschel bottle Stem Length: 165 mm 3 Pack of 5
10637615 For model MF29/3/250 Dreschel bottle Stem Length: 165 mm 1 Each
11362065 For model MF29/3/500 Dreschel bottle Stem length: 217 mm 2 Each
11332065 For model MF29/3/125 Dreschel bottle Stem Length: 132 mm 1 Each
11342065 For model MF29/3/500 Dreschel bottle Stem length: 217 mm 1 Each
11322065 For model MF29/3/500 Dreschel bottle Stem length: 217 mm 0 Each
11312065 For model MF29/3/250 Dreschel bottle Stem Length: 165 mm 0 Each
11302065 For model MF29/3/125 Dreschel bottle Stem Length: 132 mm 0 Each
15547940 For model MF29/23/250 Dreschel bottle Stem Length: 165 mm 2 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15557940 Supplier Quickfit Supplier No. MF48/33/250
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
  • Complies with BS 2461
  • Sintered dip tube disperses gas into bubbles for greater efficiency
  • Choice of sinter porosities to achieve fine bubble size
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance BS 2461
Description Stem Length: 165 mm
Material Borosilicate Glass
For Use With (Equipment) For model MF29/3/250 Dreschel bottle
Product Type Bottle Heads
Porosity 3