Learn More
Pocket for Quickfit™ Still Heads
Product Code. 12008493
Change view
Click to view available options
Length (Metric):
45 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12008493 45 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12008493 Supplier Quickfit Supplier No. SH4A
  • ONLINE EXCLUSIVE PRICE
    40.16€
    32.12€ / Pack of 1
    Save 8.04€
    20% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For incorporating thermometers into Quickfit™ jointed glassware assemblies
  • For incorporating thermometers into Quickfit™ jointed-ware assemblies
  • Pocket fits most Quickfit™ still heads
Specifications
For Use With (Application) Incorporating thermometers into Quickfit™ jointed-ware assemblies
Length (Metric) 45 mm
Material Borosilicate Glass
Product Type Pocket Thermometer