Learn More
Quickfit™ Three-Neck Round-Bottom Glass Flask
Product Code. 10228054
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Socket Joint:
14/23 (Centre), 14/23 (side)
19/26 (Centre), 14/23 (side)
19/26 (Centre), 19/26 (side)
24/29 (Centre), 14/23 (side)
24/29 (Centre), 19/26 (side)
29/32
29/32 (Centre), 14/23 (side)
29/32 (Centre), 19/26 (side)
29/32 (Centre), 29/32 (side)
34/35 (Centre), 19/26 (side)
Unit Size:
Each
Pack of 5
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Socket Joint Unit Size
10228054 1000 mL 24/29 (Centre), 19/26 (side) Each
11710259 100 mL 24/29 (Centre), 14/23 (side) Each
12020643 100 mL 24/29 (Centre), 14/23 (side) Each
11301945 100 mL 19/26 (Centre), 14/23 (side) Each
12488966 100 mL 24/29 (Centre), 19/26 (side) Each
11371955 250 mL 19/26 (Centre), 19/26 (side) Each
12408996 250 mL 24/29 (Centre), 14/23 (side) Each
12622012 250 mL 29/32 (Centre), 19/26 (side) Each
12030673 500 mL 24/29 (Centre), 19/26 (side) Each
11900895 500 mL 29/32 (Centre), 19/26 (side) Each
12493621 50 mL 14/23 (Centre), 14/23 (side) Each
12413631 50 mL 19/26 (Centre), 14/23 (side) Each
15557860 1000 mL 29/32 Pack of 5
12050653 250 mL 24/29 (Centre), 19/26 (side) Each
15577860 1000 mL 29/32 (Centre), 29/32 (side) Pack of 5
15587860 1000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15547870 250 mL 29/32 (Centre), 14/23 (side) Pack of 5
15577870 2000 mL 24/29 (Centre), 19/26 (side) Pack of 5
15527880 2000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15597880 3000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15567890 50 mL 19/26 (Centre), 19/26 (side) Pack of 5
15587890 500 mL 24/29 (Centre), 14/23 (side) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Product Code. 10228054 Supplier Quickfit Supplier No. FR1L/3S/22A
  • ONLINE EXCLUSIVE PRICE
    75.71€
    63.23€ / Pack of 1
    Save 12.48€
    16% Off
View all promotions
Estimated Shipment: 09-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
  • Fitted with three Quickfit™ precision-ground sockets
    • One vertical centre neck
    • Two angled side necks
  • Ground glass sockets are manufactured to an accuracy which surpasses the requirements of ISO 383
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
Specifications
Certifications/Compliance ISO 383
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 180 mm
No. per Case 3
Socket Joint 24/29 (Centre), 19/26 (side)
Bottom Shape Round