Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Product Code. 12403631
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Socket Joint:
14/23 (Centre), 14/23 (side)
24/29 (Centre), 14/23 (side)
24/29 (Centre), 19/26 (side)
34/35 (Centre), 19/26 (side)
Unit Size:
Each
Pack of 5
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Socket Joint unitSize
12403631 50 mL 14/23 (Centre), 14/23 (side) Each
12030643 100 mL 24/29 (Centre), 19/26 (side) Each
12478966 100 mL 24/29 (Centre), 14/23 (side) Each
12418996 250 mL 24/29 (Centre), 14/23 (side) Each
12070653 250 mL 24/29 (Centre), 19/26 (side) Each
12438986 25 mL 14/23 (Centre), 14/23 (side) Each
12050673 500 mL 24/29 (Centre), 19/26 (side) Each
15547880 2000 mL 34/35 (Centre), 19/26 (side) Pack of 5
15517860 1000 mL 24/29 (Centre), 14/23 (side) Pack of 5
15537860 1000 mL 24/29 (Centre), 19/26 (side) Pack of 5
15597890 500 mL 24/29 (Centre), 14/23 (side) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 12403631 Supplier Quickfit Supplier No. FR50/1S/1A
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
  • With highest-quality Quickfit™ ground glass socket
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Height (Metric) 89 mm
No. per Case 2
Socket Joint 14/23 (Centre), 14/23 (side)
Bottom Shape Round