missing translation for 'onlineSavingsMsg'
Learn More

Quinine hydrochloride, 100.8%, >99%, MP Biomedicals™

Product Code. 11450841 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450841 50 g 50g
11440841 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450841 Supplier MP Biomedicals Supplier No. 0215648650
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Color White
Assay Percent Range 1.008
Quantity 50 g
Formula Weight 360.9
Percent Purity >99%
Physical Form Powder
Chemical Name or Material Quinine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.