missing translation for 'onlineSavingsMsg'
Learn More

Quinine hydrochloride, MP Biomedicals™

Product Code. 11460841 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460841 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460841 Supplier MP Biomedicals Supplier No. 0215648680
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quinine Hydrochloride is an alkaloid originally isolated from the Cinchona family of South American trees. It is an antimalarial agent, anticholinergic, antihypertensive and hypoglycemic agent

Specifications

CAS 6119-47-7
Color White
Molecular Formula C20H29ClN2O4
MDL Number MFCD00151248
Quantity 100 g
Synonym quinine hcl dihydrate
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
IUPAC Name (R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol dihydrate hydrochloride
Molecular Weight (g/mol) 396.91
PubChem CID 124080947
Formula Weight 360.9
Physical Form Powder
Chemical Name or Material Quinine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.