Learn More
Olympus™ Quintuple Revolving Nosepiece, U-P5BDRE
Product Code. 12681511
Change view
Click to view available options
For Use With (Equipment):
Olympus microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12681511 Olympus microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12681511 Supplier Olympus™ Supplier No. K10017552
Attach and use up to 5 objectives with this revolving nosepiece. Olympus™ Quintuple Revolving Nosepiece, U-P5BDRE includes two centerable objective positions for darkfield objectives or brightfield objectives, a BD-M-AD adapter and a slot for a DIC slider or U-TAD plate adapter.
  • Revolving.
  • Holds up to 5 objectives.
  • Two centerable objective positions for darkfield or brightfield objectives.
  • BD-M-AD adapter.
  • Slot for DIC slider or U-TAD plate adapter.
Specifications
Type Rotary, Quintuple
Device Holds Objectives, DIC Slider
For Use With (Equipment) Olympus microscopes