Learn More
Olympus™ Quintuple Revolving Nosepiece, U-P5BDRE
Description
- Revolving.
- Holds up to 5 objectives.
- Two centerable objective positions for darkfield or brightfield objectives.
- BD-M-AD adapter.
- Slot for DIC slider or U-TAD plate adapter.
Specifications
Specifications
| Type | Rotary, Quintuple |
| Device Holds | Objectives, DIC Slider |
| For Use With (Equipment) | Olympus microscopes |