missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid, 95%, BLD Pharm™

Product Code. 30965031
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30965031 5 g 5g
30954212 100 g 100g
30950590 500 g 500g
30953296 10 g 10g
30954676 25 g 25g
30949274 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30965031 Supplier BLD Pharm Supplier No. BD134535g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 28697-17-8
Molecular Formula C11H19NO4
MDL Number MFCD00237380
Quantity 5 g
SMILES O=C([C@@H]1N(C(OC(C)(C)C)=O)CCCC1)O
Molecular Weight (g/mol) 229.27
Percent Purity 95%
Chemical Name or Material (R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.