Learn More
behr Labor-Technik™ behrotest™ R 104 S Soxhlet Extractor Unit
Product Code. 10531033
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10531033 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10531033 Supplier behr Labor-Technik™ Supplier No. B00218425
  • ONLINE EXCLUSIVE PRICE
    7973.12€
    3097.02€ / Pack of 1
    Save 4876.10€
    61% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction).
  • Infinitely variable heating regulation after the extraction cycle the compact system removes the solvent by means of a discharge valve on the extractor directly into the reservoir.
    • Includes sample package, extraction sleeves and boiling stones.
      • behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
        • With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
          • Practical holders for condenser and extraction adapter for secure placement between extractions
            • Condenser with screw connections
  • Specifications
    Product Type Extractor Unit
    Capacity (Metric) 250 mL
    Includes 4 sample positions