missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-1-(3-phenoxypropyl)quinuclidin-1-ium bromide, 98%, BLD Pharm™

Product Code. 30965532
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30965532 250 mg 250mg
30962874 25 mg 25mg
30959730 100 mg 100mg
30957628 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30965532 Supplier BLD Pharm Supplier No. BD112529250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 320345-99-1
Molecular Formula C26H30BrNO4S2
MDL Number MFCD09954105
Quantity 250 mg
SMILES O=C(O[C@H]1C[N+]2(CCCOC3=CC=CC=C3)CCC1CC2)C(C4=CC=CS4)(O)C5=CC=CS5.[Br-]
Molecular Weight (g/mol) 564.55
Percent Purity 98%
Chemical Name or Material (R)-3-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-1-(3-phenoxypropyl)quinuclidin-1-ium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.