missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Aminopyrrolidine dihydrochloride, 97%, Thermo Scientific™

Product Code. 11986761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11986761 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11986761 Supplier Thermo Scientific Acros Supplier No. 446050010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 116183-81-4
Melting Point 340°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Molecular Formula C4H10N2·2HCl
Quantity 1 g
Specific Rotation -1.1
Synonym 3r---3-aminopyrrolidine dihydrochloride, r---3-aminopyrrolidine dihydrochloride, r-pyrrolidin-3-amine dihydrochloride, r-3-aminopyrrolidine 2hcl, r-+-3-aminopyrrolidine 2hcl, 3r-pyrrolidin-3-amine dihydrochloride, 3r-3-pyrrolidinamine dihydrochloride, r-3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, r, pubchem5730
InChI Key NJPNCMOUEXEGBL-RZFWHQLPSA-N
SMILES C1CNCC1N.Cl.Cl
IUPAC Name (3R)-pyrrolidin-3-amine;dihydrochloride
Molecular Weight (g/mol) 159.06
PubChem CID 13500643
Formula Weight 159.06
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-3-Aminopyrrolidine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • (R)-3-Aminopyrrolidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.