missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

Product Code. 15477356
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477356 250 mg 250mg
15457356 1 g 1g
15467356 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15477356 Supplier Thermo Scientific Alfa Aesar Supplier No. H62381.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 198544-42-2 | C23H26N2O6 | 426.469 g/mol

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid is used as catalyst or pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198544-42-2
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00798648
InChI Key PKAUMAVONPSDRW-LJQANCHMSA-N
Synonym fmoc-d-dap boc-oh, n2-fmoc-n3-boc-d-2,3-diaminopropionic acid, fmoc-n3-boc-d-2,3-diaminopropionic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, fmoc-n-beta-boc-d-alpha,beta-diaminopropionicacid, 2r-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1320, fmoc-d-dpr boc-oh, fmoc-d-dapa boc-oh, fmoc-dap boc-oh hydrate
PubChem CID 17040127
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 426.46
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.