missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-5-phenylpentanoic acid, 95%, Thermo Scientific™

Product Code. p-7034372
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408557 250 mg 250mg
15498547 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408557 Supplier Thermo Scientific Alfa Aesar Supplier No. H52055.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 218608-83-4
Molecular Formula C16H23NO4
Molecular Weight (g/mol) 293.363
MDL Number MFCD01076262
InChI Key MYWZFJXOLAXENE-CYBMUJFWSA-N
Synonym r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, r-3-boc-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonyl amino-5-phenylpentanoic acid, boc-d-?-nva 5-phenyl-oh, n-boc-3-r-amino-5-phenylpentanoic acid, n-tert-butoxycarbonyl-3 r-amino-5-phenylpentanoic acid, 3r-3-tert-butoxycarbonylamino-5-phenyl-pentanoic acid, r-3-tert-butoxycarbonyl amino-5-phenylpentanoicacid, r-3-boc-amino-5-phenylpentanoic acid hplc, 3r-3-2-methylpropan-2-yl oxycarbonylamino-5-phenylpentanoic acid
PubChem CID 7021558
IUPAC Name (3R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-5-phenylpentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)CC(=O)O

Specifications

Quantity 250 mg
Formula Weight 293.36
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-5-phenylpentanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.