missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Chloro-1-phenyl-1-propanol, TRC

Product Code. 17931785
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17931785 5 g 5g
17921785 10 g 10g
17931145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931785 Supplier Toronto Research Chemicals Supplier No. TRCC3795965G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100306-33-0
Molecular Formula C9 H11 Cl O
Molecular Weight (g/mol) 170.64
Synonym (αR)-α-(2-Chloroethyl)benzenemethanol, (R)-3-Chloro-1-phenyl-1-propanol
IUPAC Name (1R)-3-chloro-1-phenylpropan-1-ol
SMILES O[C@H](CCCl)c1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H11ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9,11H,6-7H2/t9-/m1/s1
Formula Weight 170.0498
Chemical Name or Material (R)-(+)-3-Chloro-1-phenyl-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.