missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Hydroxypyrrolidine hydrochloride, 97%, Thermo Scientific™

Product Code. 15489477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489477 5 g 5g
15479477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489477 Supplier Thermo Scientific Alfa Aesar Supplier No. H52743.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-3-Hydroxypyrrolidine hydrochloride is used as a darifenacin Intermediate.

Solubility
Soluble in DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104706-47-0
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00191570
InChI Key QPMSJEFZULFYTB-PGMHMLKASA-N
Synonym r-3-hydroxypyrrolidine hydrochloride, r---3-pyrrolidinol hydrochloride, 3r-pyrrolidin-3-ol hydrochloride, r-pyrrolidin-3-ol hydrochloride, r-3-hydroxypyrrolidine hcl, r-3-pyrrolidinol hydrochloride, 3r-3-hydroxypyrrolidine hydrochloride, 3r-pyrrolidinol hydrochloride, r-+-3-pyrrolidinol hydrochloride
PubChem CID 2759336
IUPAC Name (3R)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 4450078
Solubility Information Soluble in DMSO.
Optical Rotation −7.6° (c=3.5 in Methanol)
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material (R)-3-Hydroxypyrrolidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.