missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-3-Quinuclidinol-2,2,3,6,6,7,7-d7, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | (3 R)-3-Quinuclidinol-2,2,3,6,6,7,7-D7 |
| Molecular Formula | C7 D7 H6 N O |
| Quantity | 0.01 g |
| Synonym | (3 R)-2,2,3,6,6,7,7-Heptadeuterioquinuclidin-3-ol, (R)-(-)-3-Quinuclidinol-d7, (3 R)-3-Quinuclidinol-2,2,3,6,6,7,7-D7, (3 R)-2,2,3,6,6,7,7-Heptadeuterio-1-azabicyclo[2.2.2 ]octan-3-ol, (3 R)-1-Azabicyclo[2.2.2 ]octan-3-ol-2,2,3,6,6,7,7-d7, (-)-3-Quinuclidinol-2,2,3,6,6,7,7-d7, (R)-(-)-3-Quinuclidinol-2,2,3,6,6,7,7-d7 |
| InChI Formula | InChI=1 S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9 H,1-5H2/t7-/m0/s1/i3D2,4D2,5D2,7 D |
| SMILES | [2 H]C1([2 H])CC2CC([2 H])([2 H])N1C([2 H])([2 H])[C@]2([2 H])O |
| IUPAC Name | (3 R)-2,2,3,6,6,7,7-heptadeuterio-1-azabicyclo[2.2.2 ]octan-3-ol |
| Molecular Weight (g/mol) | 134.2274 |
| Formula Weight | 134.1437 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction