missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-5 ELISpot Kit

Product Code. 18732054
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732054 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732054 Supplier R&D Systems™ Supplier No. EL205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least one 96-well plates

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.