Learn More
R&D Systems™ Human TGF-beta RII (NP_003233) VersaClone cDNA
Product Code. 18745908
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18745908 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18745908 missing translation for 'mfr' R&D Systems™ missing translation for 'supplierNo' RDC1270
Este artículo no se puede devolver. Vea la política de devoluciones
TGF-beta receptor type IIB; TGF-beta receptor type-2; TGF-beta type II receptor; TGFbeta-RII; TGFR-2; transforming growth factor beta receptor type IIC
The Human TGF-beta RII (NP_003233) VersaClone cDNA clone contains the complete ORF for TGF-beta RII, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TGF-beta RII insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Información de órdenes
Ships at ambient temperature
TRUSTED_SUSTAINABILITY
Especificaciones
Product Type cDNA
Gene hTGFBR2
Accession Number NP_003233
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1717bp
Gene ID (Entrez) 7048, 21813
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human, Mouse
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.