Learn More
R&D Systems™ Proteome Profiler Human Protease Inhibitor Array Kit
GREENER_CHOICE
Product Code. 18017631
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18017631 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18017631 Supplier R&D Systems™ Supplier No. ARY023
Rapid, sensitiv, and economical tool to simultaneously detect the relative changes of 32 protease inhibiting analytes in a single sample
The Proteome Profiler Human Protease Inhibitor Array Kit contains 4 membranes - each spotted in duplicate with 32 different protease inhibitor antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit