Learn More
R&D Systems™ Proteome Profiler Mouse Chemokine Array Kit
Description
The Proteome Profiler Mouse Chemokine Array Kit contains 4 membranes - each spotted in duplicate with 25 different chemokine antibodies.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Quantity | 1 Kit |