Learn More
R&D Systems™ Proteome Profiler Rat Adipokine Array Kit
GREENER_CHOICE
Product Code. 18057621
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18057621 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18057621 Supplier R&D Systems™ Supplier No. ARY016
The Proteome Profiler Rat Adipokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Rat Adipokine Array Kit contains 4 membranes - each spotted in duplicate with 30 different obesity-related antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit