missing translation for 'onlineSavingsMsg'
Learn More

(R)-Diclofop-methyl, TRC

Product Code. 30179929
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179929 50 mg 50mg
30180135 100 mg 100mg
30180317 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179929 Supplier Toronto Research Chemicals Supplier No. TRCD35498050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71283-65-3
Molecular Formula C16 H14 Cl2 O4
Molecular Weight (g/mol) 341.186
Synonym Propanoic acid, 2-[4-(2,4-dichlorophenoxy)phenoxy]-, methyl ester, (R)-, (+)-Diclofop-methyl, (R)-(+)-Diclofop-methyl, (R)-Dichlofop-methyl, (R)-Diclofop-methyl, Diclofop-P-methyl
IUPAC Name methyl (2R)-2-[4-(2,4-dichlorophenoxy)phenoxy]propanoate
SMILES COC(=O)[C@@H](C)Oc1ccc(Oc2ccc(Cl)cc2Cl)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14Cl2O4/c1-10(16(19)20-2)21-12-4-6-13(7-5-12)22-15-8-3-11(17)9-14(15)18/h3-10H,1-2H3/t10-/m1/s1
Formula Weight 340.0269
Chemical Name or Material Diclofop-P-methyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.