missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Connexin 62/GJA10 (NP_115991) VersaClone cDNA
Beskrivelse
The Human Connexin 62/GJA10 (NP_115991) VersaClone cDNA clone contains the complete ORF for Connexin 62/GJA10, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Connexin 62/GJA10 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature
Tekniske data
Tekniske data
| Accession Number | NP_115991 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?