missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-Desacryloyl N-Propionyl Ibrutinib, TRC

Product Code. 30011503
Change view
Click to view available options
Quantity:
10 mg
1 g
Unit Size:
10mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011503 10 mg 10mg
30004936 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011503 Supplier Toronto Research Chemicals Supplier No. TRCD28816510MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1839099-22-7
Molecular Formula C25 H26 N6 O2
Molecular Weight (g/mol) 442.51
Synonym 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-1-propanone
IUPAC Name 1-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidyl]propan-1-one
SMILES CCC(=O)N1CCC[C@H](C1)n2nc(c3ccc(Oc4ccccc4)cc3)c5c(N)ncnc25

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H26N6O2/c1-2-21(32)30-14-6-7-18(15-30)31-25-22(24(26)27-16-28-25)23(29-31)17-10-12-20(13-11-17)33-19-8-4-3-5-9-19/h3-5,8-13,16,18H,2,6-7,14-15H2,1H3,(H2,26,27,28)/t18-/m1/s1
Formula Weight 442.2117
Chemical Name or Material (R)-N-Desacryloyl N-Propionyl Ibrutinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.