missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Propylene Oxide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | (R)-(+)-Propylene Oxide |
| CAS | 15448-47-2 |
| Molecular Formula | C3H6O |
| Quantity | 25 g |
| Synonym | (+)-(R)-Propylene Oxide, (+)-Methyloxirane, (+)-Propylene Oxide, (2R)-2-Methyloxirane, (2R)-Methyloxirane, (R)-(+)-1,2-Epoxypropane, (R)-(+)-1,2-Epoxypropane, (R)-(+)-Methyloxirane, (R)-1,2-Epoxypropane, (R)-2-Methyloxirane, (R)-Methyloxirane, (R)-Propenoxide, (R)-Propylene Oxide, d-Propylene Oxide |
| InChI Formula | InChI=1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3 |
| SMILES | C[C@@H]1CO1 |
| IUPAC Name | (R)-2-methyloxirane |
| Molecular Weight (g/mol) | 58.08 |
| Formula Weight | 58.04 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction