missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene Oxide, TRC

Product Code. 30424568
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30424568

Brand: Toronto Research Chemicals TRCP8352601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(+)-Propylene Oxide
CAS 15448-47-2
Molecular Formula C3H6O
Quantity 1 g
Synonym (+)-(R)-Propylene Oxide, (+)-Methyloxirane, (+)-Propylene Oxide, (2R)-2-Methyloxirane, (2R)-Methyloxirane, (R)-(+)-1,2-Epoxypropane, (R)-(+)-1,2-Epoxypropane, (R)-(+)-Methyloxirane, (R)-1,2-Epoxypropane, (R)-2-Methyloxirane, (R)-Methyloxirane, (R)-Propenoxide, (R)-Propylene Oxide, d-Propylene Oxide
InChI Formula InChI=1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3
SMILES C[C@@H]1CO1
IUPAC Name (R)-2-methyloxirane
Molecular Weight (g/mol) 58.08
Formula Weight 58.04
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.