Learn More
R&D Systems™ Quantikine Immunoassay Control Group 10
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | High Sensitivity IL-1 beta, IL-4, IL-7, IL-8, IL-10, IL-12 p70, FGF basic, GM-CSF, TNF-alpha |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |