missing translation for 'onlineSavingsMsg'
Learn More

(R,S)-Alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic Acid, TRC

Product Code. 30138639
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138639 50 mg 50mg
30137431 100 mg 100mg
30004906 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138639 Supplier Toronto Research Chemicals Supplier No. TRCA61150050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77521-29-0
Molecular Formula C7 H10 N2 O4
Molecular Weight (g/mol) 186.17
Synonym alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, α-Amino-2,3-dihydro-5-methyl-3-oxo-4-isoxazolepropanoic acid, (R,S)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, (RS)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, (±)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 2-Amino-3-(3-hydroxy-5-methyl-1,2-oxazol-4-yl)propanoic acid, 2-Amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid, 2-Amino-3-(5-methyl-3-oxo-2,3-dihydroisoxazol-4-yl)propanoic acid, AMPA, AMPA (pharmaceutical), D,L-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, dl-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, α-Amino-2,3-dihydro-5-methyl-3-oxoisoxazole-4-propionic acid, α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionate, α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid, γ-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid
IUPAC Name 2-amino-3-(3-hydroxy-5-methyl-1,2-oxazol-4-yl)propanoic acid
SMILES Cc1onc(O)c1CC(N)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H10N2O4/c1-3-4(6(10)9-13-3)2-5(8)7(11)12/h5H,2,8H2,1H3,(H,9,10)(H,11,12)
Formula Weight 186.0641
Chemical Name or Material (r,s)-a-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.