Learn More
(R,S)-Anatabine-2,4,5,6-d4, TRC
Product Code. 30339036
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339036 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30339036 Supplier Toronto Research Chemicals Supplier No. TRCA6375051MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R,S)-Anatabine-2,4,5,6-d4
CAS 1020719-11-2
Purity Grade Notes HPLC
Molecular Formula C10 2H4 H8 N2
Quantity 1 mg
Synonym (RS)-Anatabine-2,4,5,6-d4, rac-Anatabine-D4 (pyridine-D4)
InChI Formula InChI=1S/C10H12N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h1-4,6,8,10,12H,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CC=CCN2)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-(1,2,3,6-tetrahydropyridin-2-yl)pyridine
Molecular Weight (g/mol) 164.24
Formula Weight 164.1252
Percent Purity >95