missing translation for 'onlineSavingsMsg'
Learn More

(R/S)-delta-Hexalactone, TRC

Product Code. 30396540
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30396540 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396540 Supplier Toronto Research Chemicals Supplier No. TRCH294195250G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material delta-Hexalactone
CAS 823-22-3
Molecular Formula C6 H10 O2
Quantity 250 g
Synonym 2H-Pyran-2-one, tetrahydro-6-methyl-, Hexanoic acid, 5-hydroxy-, lactone (6CI,7CI), Tetrahydro-6-methyl-2H-pyran-2-one, (RS)-Δ-Hexalactone, (±)-5-Hexanolide, (±)-Δ-Hexalactone, 5-Hexanolide, 5-Hydroxyhexanoic acid lactone, 6-Methyl-2H-tetrahydropyran-2-one, 6-Methyl-Δ-valerolactone, 6-Methyltetrahydropyran-2-one, 6-Methylvalerolactone, Hexanoic acid, 5-hydroxy-, Δ-lactone, NSC 134774, NSC 32863, Δ-Caprolactone, Δ-Hexalactone, Δ-Hexanolactone, Δ-Hexanolide, Δ-Methyl-Δ-valerolactone
InChI Formula InChI=1S/C6H10O2/c1-5-3-2-4-6(7)8-5/h5H,2-4H2,1H3
SMILES CC1CCCC(=O)O1
IUPAC Name 6-methyloxan-2-one
Molecular Weight (g/mol) 114.14
Formula Weight 114.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.