Learn More
(R,S)-N-Nitroso Anatabine-2,4,5,6-d4 (10 ug/ml in Acetonitrile), TRC
Product Code. 30407056
Change view
Click to view available options
Quantity:
1x15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407056 1x15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30407056 Supplier Toronto Research Chemicals Supplier No. TRCKIT07741X15ML
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (RS)-N-Nitrosoanabasine pyridinyl-d4
CAS 1020719-68-9
Molecular Formula C10 2H4 H9 N3 O
Quantity 1x15 mL
Synonym Pyridine-2,3,4,6-d4, 5-(1-nitroso-2-piperidinyl)-, 5-(1-Nitroso-2-piperidinyl)pyridine-2,3,4,6-d4, (R,S)-N-Nitroso Anabasine-d4, (RS)-N-Nitrosoanabasine D4 (pyridinyl D4), (RS)-N-Nitrosoanabasine D4 (pyridinyl D4)
InChI Formula InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/i3D,4D,6D,8D
SMILES [2H]c1nc([2H])c(C2CCCCN2N=O)c([2H])c1[2H]
IUPAC Name 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine
Molecular Weight (g/mol) 195.25
Formula Weight 195.131
Concentration or Composition (by Analyte or Components) 10 μg/mL