Learn More
R-Spondin 2 Antibody (CL14321) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26851AF350
Product Code. 30297470
  • 791.00€ / 0.10mL
Description
R-Spondin 2 Monoclonal antibody specifically detects R-Spondin 2 in Human,Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
R-Spondin 2
Monoclonal
Alexa Fluor 350
CRISTIN2, hRspo2, MGC35555, MGC43342, Roof plate-specific spondin-2, R-spondin 2 homolog (Xenopus laevis), R-spondin-2
This antibody was developed using a synthetic peptide derived from Q6UXX9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse
Purified
Immunohistochemistry
CL14321
50mM Sodium Borate
Mouse
Protein A purified
RUO
340419
Store at 4⪚C in the dark.