missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | RAB11B |
|---|---|
| Clone | 1G7D6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236816
|
Proteintech
67780-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234904
|
Proteintech
67780-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAB11B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB11B. This antibody recognizes human, mouse, rat antigen. The RAB11B antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| RAB11B | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 9230 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1G7D6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| H-YPT3; MGC133246 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel