missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB11B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19742-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to RAB11B. The antibody has no cross reaction to RAB11A.
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 11 is a known marker for protein trafficking, sorting, and recycling in the endosomal pathway.Specifications
| RAB11B | |
| Polyclonal | |
| Unconjugated | |
| RAB11B | |
| GTP binding protein YPT3, H YPT3, RAB11B, Ras related protein Rab 11B, YPT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19326, 79434, 9230 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35509, P46638, Q15907 | |
| RAB11B | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction