Learn More
RAB11FIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18136-1-AP-150UL
This item is not returnable.
View return policy
Description
RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.Specifications
| RAB11FIP2 | |
| Polyclonal | |
| Unconjugated | |
| RAB11FIP2 | |
| KIAA0941, nRip11, Rab11 FIP2, RAB11FIP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22841, 308003, 74998 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| G3XA57, Q7L804 | |
| Rab11fip2 | |
| RAB11FIP2 Fusion Protein Ag12736 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |