missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570799
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 86%; Zebrafish: 100%.
RAB19 gene ontology annotations related to this gene include endoplasmic reticulum to Golgi vesicle-mediated transport; intracellular protein transport; Rab protein signal transduction.
Specifications
| RAB19 | |
| Polyclonal | |
| Unconjugated | |
| RAB19 | |
| GTP-binding protein RAB19B; hypothetical protein LOC506911; Rab19; RAB19, member RAS oncogene family; RAB19B; Ras-related protein Rab-19; ras-related protein Rab-19-like; zmp:0000001111 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 401409 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A4D1S5 | |
| RAB19 | |
| synthetic peptide directed towards the N-terminal region of human RAB19. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction