Learn More
RAB20 Monoclonal Antibody (OTI1F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505094S
Description
RAB20 Monoclonal antibody specifically detects RAB20 in Human samples. It is validated for Western BlotSpecifications
| RAB20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9ULV8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23624 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1F1 | |
| Unconjugated | |
| Cblc | |
| AA536966; D8Ertd350e; RAB20; RAB20, member RAS oncogene family; ras-related protein Rab-20 | |
| Human recombinant protein fragment corresponding to amino acids 244-474 of human CBLC produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.