missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB27A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544580
This item is not returnable.
View return policy
Description
Peptide sequence: SQAIEMLLDL IMKRMERCVD KSWIPEGVVR SNGHASTDQL SEEKEKGACG Sequence homology: Cow: 79%; Goat: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Rat: 93%; Sheep: 79%.
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.
Specifications
| RAB27A | |
| Polyclonal | |
| Unconjugated | |
| RAB27A | |
| 2210402C08Rik; 2410003M20Rik; 4933437C11Rik; ash; ashen; GS2; GTP-binding protein Ram; HsT18676; low Mr GTP-binding protein; mutant Ras-related protein Rab-27A; RAB27; rab-27; Rab27a; RAB27A, member RAS oncogene family; ram; Ram p25; ras-related protein Rab-27A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5873 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P51159 | |
| RAB27A | |
| Synthetic peptide directed towards the middle region of human RAB27A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction